Skip to Content
Merck

BDPH9BCDFD61

Bide Pharmatech

1,4-Diiodobenzene

Synonym(s):

1,4-diiodobenzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Product BDPH9BCDFD61 is not currently sold in your country.

About This Item

Linear Formula:
C6H4I2
CAS Number:
Molecular Weight:
329.91
MDL number:
Product BDPH9BCDFD61 is not currently sold in your country.

Inchi Code

1S/C6H4I2/c7-5-1-2-6(8)4-3-5/h1-4H

Inchi Key

LFMWZTSOMGDDJU-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, sealed in dry, room temperature

Country of Origin

CN

Physical Form

Form: Crystal - Powder

Melting Point

131-133C

Looking for similar products? Visit Product Comparison Guide

hstatements

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library