Skip to Content
Merck

BDPH9BCDFD69

Bide Pharmatech

1-(2,2,2-Trifluoroethyl)piperazine

Synonym(s):

1-(2,2,2-trifluoroethyl)piperazine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H11N2F3
CAS Number:
Molecular Weight:
168.16
MDL number:
Product BDPH9BCDFD69 is not currently sold in your country.


Inchi Code

1S/C6H11F3N2/c7-6(8,9)5-11-3-1-10-2-4-11/h10H,1-5H2

Inchi Key

GFPOCAYEKCWFPD-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Colorless liquid



Still not finding the right product?

Explore all of our products under 1-(2,2,2-Trifluoroethyl)piperazine


hstatements

signalword

Danger



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library