Skip to Content
Merck

BDPH9BCE00A0

Bide Pharmatech

7-tert-Butyl 2-ethyl 5,6-dihydroimidazo[1,2-a]pyrazine-2,7(8H)-dicarboxylate

Synonym(s):

7-tert-butyl 2-ethyl 5,6-dihydroimidazo[1,2-a]pyrazine-2,7(8H)-dicarboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Product BDPH9BCE00A0 is not currently sold in your country.

About This Item

Linear Formula:
C14H21O4N3
CAS Number:
Molecular Weight:
295.34
MDL number:
Product BDPH9BCE00A0 is not currently sold in your country.

Inchi Code

1S/C14H21N3O4/c1-5-20-12(18)10-8-16-6-7-17(9-11(16)15-10)13(19)21-14(2,3)4/h8H,5-7,9H2,1-4H3

Inchi Key

UQNMISYKKAQDDT-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

White to Yellow Solid

Looking for similar products? Visit Product Comparison Guide

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library