Skip to Content

BDPH9BCE17CB

Bide Pharmatech

4-(4-Iodophenyl)pyridine

Synonym(s):

4-(4-iodophenyl)pyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H8N1I1
CAS Number:
Molecular Weight:
281.1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H8IN/c12-11-3-1-9(2-4-11)10-5-7-13-8-6-10/h1-8H

Inchi Key

UDXJNRYHGQJHGL-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 4-(4-Iodophenyl)pyridine


hstatements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library