Skip to Content
Merck

BDPH9C01FBBC

Bide Pharmatech

(R)-(4-Benzylmorpholin-2-yl)methanamine

Synonym(s):

(R)-(4-benzylmorpholin-2-yl)methanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H18O1N2
CAS Number:
Molecular Weight:
206.29
MDL number:
Product BDPH9C01FBBC is not currently sold in your country.


Inchi Code

1S/C12H18N2O/c13-8-12-10-14(6-7-15-12)9-11-4-2-1-3-5-11/h1-5,12H,6-10,13H2/t12-/m1/s1

Inchi Key

CKZVBXBEDDAEFE-GFCCVEGCSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library