Skip to Content
Merck

BDPH9C020A30

Bide Pharmatech

Hexa-2,4-diyne-1,6-diyl dibenzoate

Synonym(s):

hexa-2,4-diyne-1,6-diyl dibenzoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C20H14O4/c21-19(17-11-5-3-6-12-17)23-15-9-1-2-10-16-24-20(22)18-13-7-4-8-14-18/h3-8,11-14H,15-16H2

Inchi Key

JCZJRGIBGWSWTP-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?

Explore all of our products under Hexa-2,4-diyne-1,6-diyl dibenzoate




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library