Skip to Content
Merck

BDPH9C020C19

Bide Pharmatech

Ethyl 2-(trans-4-aminocyclohexyl)acetate

Synonym(s):

ethyl (4-aminocyclohexyl)acetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Product BDPH9C020C19 is not currently sold in your country.

About This Item

Linear Formula:
C10H19O2N1
CAS Number:
Molecular Weight:
185.27
MDL number:
Product BDPH9C020C19 is not currently sold in your country.

Inchi Code

1S/C10H19NO2/c1-2-13-10(12)7-8-3-5-9(11)6-4-8/h8-9H,2-7,11H2,1H3/t8-,9-

Inchi Key

SBVCDGKSVSHGFL-KYZUINATSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Colorless or white to yellow liquid or low-melting solid

Looking for similar products? Visit Product Comparison Guide

hstatements

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library