Skip to Content
Merck

BDPH9C021D04

Bide Pharmatech

rel-(1R,4E,pS)-Cyclooct-4-enol

Synonym(s):

4-cycloocten-1-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H14O1
CAS Number:
Molecular Weight:
126.2
MDL number:
Product BDPH9C021D04 is not currently sold in your country.


Inchi Code

1S/C8H14O/c9-8-6-4-2-1-3-5-7-8/h1-2,8-9H,3-7H2/b2-1-

Inchi Key

UCPDHOTYYDHPEN-UPHRSURJSA-N

Shipping Temperature

Normal

Purity

95%14

Country of Origin

CN

Physical Form

Solid or Semi-solid or liquid or lump

Storage temp.

Inert atmosphere, store in freezer, under -20°C



Still not finding the right product?

Explore all of our products under rel-(1R,4E,pS)-Cyclooct-4-enol


hstatements

signalword

Danger

pictograms

Corrosion


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library