Skip to Content
Merck

JWPH580E1369

J & W Pharmlab LLC

4-(tert-butoxycarbonylamino)picolinic acid

Sign In to View Organizational & Contract Pricing

Select a Size

Product JWPH580E1369 is not currently sold in your country.

About This Item

CAS Number:
Molecular Weight:
238.24
MDL number:
Product JWPH580E1369 is not currently sold in your country.

Iupac Name

4-[(tert-butoxycarbonyl)amino]-2-pyridinecarboxylic acid

Inchi Code

1S/C11H14N2O4/c1-11(2,3)17-10(16)13-7-4-5-12-8(6-7)9(14)15/h4-6H,1-3H3,(H,14,15)(H,12,13,16)

InChI key

NYNGMBRXAOOPEY-UHFFFAOYSA-N

Purity

96%

Country of Origin

US

Physical Form

Off White Solid

Product Link

https://www.jwpharmlab.com/home/product/69R1375

MSDS

https://www.jwpharmlab.com/home/product/69R1375


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library