Sign In to View Organizational & Contract Pricing
Select a Size
Product LEYH9ACFB92F is not currently sold in your country.
Product LEYH9ACFB92F is not currently sold in your country.
Iupac Name
(3,4-dihydroxyphenyl)acetonitrile
Inchi Code
1S/C8H7NO2/c9-4-3-6-1-2-7(10)8(11)5-6/h1-2,5,10-11H,3H2
InChI key
IXDSDBYUZFMVBW-UHFFFAOYSA-N
storage temp.
2-8C, stored under nitrogen
Purity
98%
Country of Origin
CN
Shipping Temperature
2-8C
Physical Form
Solid
Product Link
https://www.leyan.com/1126-62-1.html
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.leyan.com/1126-62-1.html
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
