Skip to Content
Merck

PG9H9BC7B2D9

PrecisePEG

FT-FAPI-12_9 

Synonym(s):

(S)-N-(2-(2-cyano-4,4-difluoropyrrolidin-1-yl)-2-oxoethyl)-6-(methyl(3-(piperazin-1-yl)propyl)amino)quinoline-4-carbimidic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Product PG9H9BC7B2D9 is not currently sold in your country.


Inchi Code

1S/C25H31F2N7O2/c1-32(9-2-10-33-11-7-29-8-12-33)18-3-4-22-21(13-18)20(5-6-30-22)24(36)31-16-23(35)34-17-25(26,27)14-19(34)15-28/h3-6,13,19,29H,2,7-12,14,16-17H2,1H3,(H,31,36)/t19-/m0/s1

Inchi Key

KAZYDEMPAXZEJT-IBGZPJMESA-N

Shipping Temperature

ambient

Purity

96%

Storage temp.

-20

Country of Origin

CN

Physical Form

Red



hstatements

pcodes



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library