Skip to Content
Merck

RIE156324314

Rieke Metals, Inc.

4'-piperidinomethyl-3-trifluoromethylbenzophenone

Synonym(s):

[4-(1-piperidinylmethyl)phenyl][3-(trifluoromethyl)phenyl]methanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
347.38
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C20H20F3NO/c21-20(22,23)18-6-4-5-17(13-18)19(25)16-9-7-15(8-10-16)14-24-11-2-1-3-12-24/h4-10,13H,1-3,11-12,14H2

Inchi Key

ZDDYZBNTBMCOCX-UHFFFAOYSA-N

Purity

97%

Country of Origin

US



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library