Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
12171500
Pricing and availability is not currently available.
storage temp.
−20°C
Application
The Nerve Terminal Stain Kits are used to detect and study synaptic vesicle recycling in neuronal synapses and neuromuscular junctions. Although each kit employs a different amphiphilic styryl pyridinium dye to visualize the vesicles, the tissue preparation, reagents required, and staining procedures are identical for each. The dyes fluoresce intensely inside plasma membranes but only minimally in aqueous environments.
When neurons are actively releasing neurotransmitters, dye is incorporated into recycled vesicles at the presynaptic terminal. The dye-containing vesicles are not susceptible to subsequent wash steps that remove the excess dye from the outer leaflet of the plasma membrane. Thus only active presynaptic terminals are labeled. Advasep-7 further reduces background by encapsulating the probes, preventing stain from accumulating interstitially.
When neurons are actively releasing neurotransmitters, dye is incorporated into recycled vesicles at the presynaptic terminal. The dye-containing vesicles are not susceptible to subsequent wash steps that remove the excess dye from the outer leaflet of the plasma membrane. Thus only active presynaptic terminals are labeled. Advasep-7 further reduces background by encapsulating the probes, preventing stain from accumulating interstitially.
Other Notes
NTS-I utilizes SynaptoGreen™ C4 and the sulfonated β-cyclodextrin ADVASEP-7.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.