Skip to Content
Merck

B3399

Biotin

suitable for plant cell culture, ≥99% (TLC)

Synonym(s):

D-Biotin, Bios II, Coenzyme R, Vitamin B7, Vitamin H

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H16N2O3S
CAS Number:
Molecular Weight:
244.31
UNSPSC Code:
12352201
PubChem Substance ID:
eCl@ss:
34058011
EC Number:
200-399-3
Beilstein/REAXYS Number:
86838
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99% (TLC)

form

powder

mol wt

Mw 244.31 g/mol

technique(s)

cell culture | plant: suitable

color

white to off-white

mp

231-233 °C (lit.)

solubility

NH4OH: soluble 50 mg/mL, clear, colorless to faintly yellow (2 M Ammonium hydroxide)

storage temp.

2-8°C

SMILES string

[H][C@]12CS[C@@H](CCCCC(O)=O)[C@@]1([H])NC(=O)N2

InChI

1S/C10H16N2O3S/c13-8(14)4-2-1-3-7-9-6(5-16-7)11-10(15)12-9/h6-7,9H,1-5H2,(H,13,14)(H2,11,12,15)/t6-,7-,9-/m0/s1

InChI key

YBJHBAHKTGYVGT-ZKWXMUAHSA-N



Still not finding the right product?

Explore all of our products under Biotin


Storage Class

13 - Non Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library