Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41106514
eCl@ss:
32040503
Biological source:
rat brain
Relevant disease(s):
cancer
Growth mode:
Adherent
Karyotype:
modal No varied with passage see reference
Morphology:
fibroblastic
Manufacturer/tradename:
ECACC
Product Name
C6-2-3, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
biological source
rat brain
manufacturer/tradename
ECACC
packaging
tube of 5 μg 08062528-DNA-5UG, pkg of vial of cells 08062528-1VL
growth mode
Adherent
karyotype
modal No varied with passage see reference
morphology
fibroblastic
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Application
electrophysiology
Biochem/physiol Actions
A pseudotetraploid call line generated by fusion of 8-bromodeoxyuridine-resistant mutant rat glioma cell line C6-BU-1 and the 6-thioguanine-resistant mutant rat glioma cell line C6-TG-1.
Rat glioma x rat glioma hybrid
Preparation Note
DMEM + 2 mM glutamine + 10% Foetal Bovine Serum (FBS) + HAT (0.1 mM hypoxanthine, 10 μM aminopterin and 16 μM thymidine) Cells can be cultured without HAT provided they were cultured in between in HT medium for 3-4 days
Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.05% trypsin/EDTA; 5% CO2; 37°C
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). No licensing agreement is required when either this cell line or the DNA extracted from it are used for internal research purposes only. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to culturecollections@ukhsa.gov.uk.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.