Skip to Content
Merck

54SCC2 Mouse x Rat Hybridoma Nerve, ECACC 08062560

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106514
eCl@ss:
32160701
Biological source:
Mouse x Rat Hybridoma nerve
Relevant disease(s):
cancer
Growth mode:
Adherent
Karyotype:
Modal No. varied with passage see reference
Morphology:
oligopolar
Manufacturer/tradename:
ECACC
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

54SCC2, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

biological source

Mouse x Rat Hybridoma nerve

manufacturer/tradename

ECACC

packaging

tube of 5 μg 08062560-DNA-5UG, pkg of vial of cells 08062560-1VL

growth mode

Adherent

karyotype

Modal No. varied with passage see reference

morphology

oligopolar

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Biochem/physiol Actions

Hybrid generated by fusion of bromodeoxyuridine-resistant mutant of rat glioma cell line C6 (C6-BU-1, Sigma Catalogue number 08062563) and the 6-thioguanine-resistant mouse L cell fibroblast line A9 (Sigma Catalogue number 08062526) using inactivated Sendai virus. Cells formed from such a fusion are multipolar and extend small processes.
Rat glioma x mouse L cell fibroblast hybrid

Preparation Note

DMEM + 2 mM glutamine + 10% Foetal Bovine Serum (FBS) + HAT (0.1 mM hypoxanthine, 10 μM aminopterin and 16 μM thymidine) Cells can be cultured without HAT provided they were cultured in between in HT medium for 3-4 days
Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.05% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). No licensing agreement is required when either this cell line or the DNA extracted from it are used for internal research purposes only. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to culturecollections@ukhsa.gov.uk.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library