Skip to Content
Merck

E5031

Monoclonal Anti-ERp57 antibody produced in mouse

~2 mg/mL, clone TO-2, purified immunoglobulin, buffered aqueous solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
MDL number:
eCl@ss:
32160702
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

TO-2, monoclonal

form

buffered aqueous solution

mol wt

antigen ~57 kDa

species reactivity

human, mouse

concentration

~2 mg/mL

technique(s)

flow cytometry: suitable, immunocytochemistry: suitable, immunohistochemistry: suitable, indirect ELISA: suitable, microarray: suitable, western blot: 20-30 μg/mL using total cell extract of NS1 cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... PDIA3(2923)
mouse ... Pdia3(14827)

Immunogen

synthetic peptide corresponding to amino acids 460-478 of human ERp57.

Physical form

Solution in 0.01 M phos­­­­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library