About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
eCl@ss:
32160701
Material:
U-bottom wells, clear plate, clear wells, polypropylene , square wells
Size:
96 wells
Sterility:
non-sterile
Feature:
skirt: no, lid: no
material
U-bottom wells, clear plate, clear wells, polypropylene , square wells
sterility
non-sterile
feature
skirt: no, lid: no
packaging
case of 50 ea (10 bags of 5ea)
manufacturer/tradename
MTC™ Bio D3096-25
parameter
4,000 × g speed
capacity
1.2 mL
size
96 wells
well volume
1.2 mL
General description
OptiWell plates come in 24, 48, 96 or 384 format for virtually every research lab, HTS application and automated/robotic platform. These plates are useful for bacterial cultures, centrifugation (up to 4000 x g) and general sample storage. Plates are produced/packaged in a Class 100,000 clean room and are ANSI / SLAS 1-2004 compliant. Several plates are sterilized by gamma irradiation and all are certified non-pyrogenic, RNase/DNase free.
Features and Benefits
- For automated research platforms / robotic equipment
- Produced/packaged in a Class 100,000 clean room
- ANSI / SLAS 1-2004 compliant
- Minimal binding / liquid retention
- Black printed alpha-numerics
- Virgin, medical-grade polypropylene
- Sterilized by gamma irradiation
- Certified non-pyrogenic, RNase/DNase free
Legal Information
MTC is a trademark of MTC Bio
OptiWell is a trademark of MTC Bio
Still not finding the right product?
Explore all of our products under MTC™ Bio OptiWell™ Deep Well Plate
Choose from one of the most recent versions:
Already Own This Product?