Skip to Content
Merck

PROTPR

ProteoPrep® Protein Precipitation Kit

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

shelf life

≥1 yr (when stored at room temperature)

Application

The ProteoPrep Protein Precipitation Kit facilitates the concentration and purification of proteins. Once resuspended in a diluent of your choice, these proteins can be characterized through conventional protein assays, electrophoresis, mass spectrometry, or any number of applications.

Features and Benefits

  • Ready-to-use reagents save you time and ensure consistency
  • Room-temperature storage saves you precious cooler space
  • A comprehensive technical bulletin provides an optimized protocol

Legal Information

ProteoPrep is a registered trademark of Merck KGaA, Darmstadt, Germany

Kit Components Only

Product No.
Description

  • Trichloroacetic Acid 100 %

  • Deoxycholate .002 %

  • Wash Solution 25 %

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bryan M Ham et al.
Analytical and bioanalytical chemistry, 387(3), 889-900 (2007-01-11)
A study of rabbit tear protein expression in a dry eye rabbit model was performed to determine if a pattern in expressed proteins could be identified. The uniqueness of the model allows the comparison of normal (control) eye tear protein
Kaori Iida et al.
BMC cell biology, 8, 38-38 (2007-08-31)
Deficiency of the PERK eIF2 alpha kinase in humans and mice results in postnatal exocrine pancreatic atrophy as well as severe growth and metabolic anomalies in other organs and tissues. To determine if the exocrine pancreatic atrophy is due to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service