Skip to Content
Merck

SRP0331

FES active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥90% (SDS-PAGE)

Synonym(s):

FPS, feline sarcoma oncogene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
NACRES:
NA.32
UNSPSC Code:
12352202
eCl@ss:
32150313
Assay:
≥90% (SDS-PAGE)
Biological source:
human
Recombinant:
expressed in baculovirus infected Sf9 cells
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

125 kDa

packaging

pkg of 10 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... FES(2242)

General description

Human feline sarcoma oncogene (FES), (GenBank Accession No. NM_002005), amino acids 2-822 (end) with N-terminal His-tag and N-terminal Flag-tag, MW=95.3 kDa, expressed in Sf9 insect cells via a Baculovirus expression system.


Storage Class

10 - Combustible liquids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library