Sign In to View Organizational & Contract Pricing
Select a Size
About This Item
Empirical Formula (Hill Notation):
C12H12O2
CAS Number:
Molecular Weight:
188.22
MDL number:
UNSPSC Code:
41116107
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥90.0% (HPLC)
manufacturer/tradename
PhytoLab
technique(s)
HPLC: suitable
refractive index
n20/D 1.577 (lit.)
bp
139-142 °C/5 mmHg (lit.)
density
1.103 g/mL at 25 °C (lit.)
application(s)
food and beverages
pharmaceutical
format
neat
storage temp.
2-8°C
SMILES string
CCC\C=C1/OC(=O)c2ccccc12
InChI
1S/C12H12O2/c1-2-3-8-11-9-6-4-5-7-10(9)12(13)14-11/h4-8H,2-3H2,1H3/b11-8-
InChI key
WMBOCUXXNSOQHM-FLIBITNWSA-N
Looking for similar products? Visit Product Comparison Guide
General description
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
Packaging
This reference substance will be packed in micro-volume insert vials.
Legal Information
phyproof is a registered trademark of PhytoLab GmbH & Co. KG
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service