
Malt Extract Agar

Synonym(s):
Malt-based culture medium
Products
Product Category
Brand
Color
Physical Form
Product Line
Capacity (mL)
Diameter (mm)
Volume (ml)
Quality Segment
Application
Sterility
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing