Merck
  • Cell cycle-dependent phosphorylation and ubiquitination of a G protein alpha subunit.
Cell cycle-dependent phosphorylation and ubiquitination of a G protein alpha subunit.
The Journal of biological chemistry (2011-04-28)
Matthew P Torres, Sarah T Clement, Steven D Cappell, Henrik G Dohlman
ABSTRACT
A diverse array of external stimuli, including most hormones and neurotransmitters, bind to cell surface receptors that activate G proteins. Mating pheromones in yeast Saccharomyces cerevisiae activate G protein-coupled receptors and initiate events leading to cell cycle arrest in G(1) phase. Here, we show that the Gα subunit (Gpa1) is phosphorylated and ubiquitinated in response to changes in the cell cycle. We systematically screened 109 gene deletion strains representing the non-essential yeast kinome and identified a single kinase gene, ELM1, as necessary and sufficient for Gpa1 phosphorylation. Elm1 is expressed in a cell cycle-dependent manner, primarily at S and G(2)/M. Accordingly, phosphorylation of Gpa1 in G(2)/M phase leads to polyubiquitination in G(1) phase. These findings demonstrate that Gpa1 is dynamically regulated. More broadly, they reveal how G proteins can simultaneously regulate, and become regulated by, progression through the cell cycle.
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Hydroxyurea, 98%, powder