Merck
  • Binding Characteristics and Analgesic Effects of Mirogabalin, a Novel Ligand for the α2δ Subunit of Voltage-Gated Calcium Channels.
Binding Characteristics and Analgesic Effects of Mirogabalin, a Novel Ligand for the α2δ Subunit of Voltage-Gated Calcium Channels.
The Journal of pharmacology and experimental therapeutics (2018-03-23)
Yuki Domon, Naohisa Arakawa, Tatsuya Inoue, Fumihiko Matsuda, Makoto Takahashi, Naotoshi Yamamura, Kiyonori Kai, Yutaka Kitano
ABSTRACT
Mirogabalin ([(1R,5S,6S)-6-(aminomethyl)-3-ethylbicyclo[3.2.0]hept-3-en-6-yl]acetic acid), a novel ligand for the α2δ subunit of voltage-gated calcium channels, is being developed to treat pain associated with diabetic peripheral neuropathy and postherpetic neuralgia. In the present study, we investigated the in vitro binding characteristics and in vivo analgesic effects of mirogabalin compared with those of pregabalin, a standard α2δ ligand. Mirogabalin showed potent and selective binding affinities for the α2δ subunits, while having no effects on 186 off-target proteins. Similar to pregabalin, mirogabalin did not show clear subtype selectivity (α2δ-1 vs. α2δ-2) or species differences (human vs. rat). However, in contrast to pregabalin, mirogabalin showed greater binding affinities for human α2δ-1, human α2δ-2, rat α2δ-1, and rat α2δ-2 subunits; further, it had a slower dissociation rate for the α2δ-1 subunit than the α2δ-2 subunit. Additionally, in experimental neuropathic pain models, partial sciatic nerve ligation rats and streptozotocin-induced diabetic rats, mirogabalin showed more potent and longer lasting analgesic effects. In safety pharmacological evaluations, mirogabalin and pregabalin inhibited rota-rod performance and locomotor activity in rats; however, the safety indices of mirogabalin were superior to those of pregabalin. In conclusion, mirogabalin shows potent and selective binding affinities for the human and rat α2δ subunits, and slower dissociation rates for the α2δ-1 subunit than the α2δ-2 subunit. It shows potent and long-lasting analgesic effects in rat models of neuropathic pain, and wider safety margins for side effects of the central nervous system. These properties of mirogabalin can be associated with its unique binding characteristics.
Sigma-Aldrich
Anti-Calcium Channel (α2/δ-1 Subunit) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
Pricing and availability is not currently available.