Merck
  • m6A RNA Methylation Maintains Hematopoietic Stem Cell Identity and Symmetric Commitment.
m6A RNA Methylation Maintains Hematopoietic Stem Cell Identity and Symmetric Commitment.
Cell reports (2019-08-15)
Yuanming Cheng, Hanzhi Luo, Franco Izzo, Brian F Pickering, Diu Nguyen, Robert Myers, Alexandra Schurer, Saroj Gourkanti, Jens C Brüning, Ly P Vu, Samie R Jaffrey, Dan A Landau, Michael G Kharas
ABSTRACT
Stem cells balance cellular fates through asymmetric and symmetric divisions in order to self-renew or to generate downstream progenitors. Symmetric commitment divisions in stem cells are required for rapid regeneration during tissue damage and stress. The control of symmetric commitment remains poorly defined. Using single-cell RNA sequencing (scRNA-seq) in combination with transcriptomic profiling of HSPCs (hematopoietic stem and progenitor cells) from control and m6A methyltransferase Mettl3 conditional knockout mice, we found that m6A-deficient hematopoietic stem cells (HSCs) fail to symmetrically differentiate. Dividing HSCs are expanded and are blocked in an intermediate state that molecularly and functionally resembles multipotent progenitors. Mechanistically, RNA methylation controls Myc mRNA abundance in differentiating HSCs. We identified MYC as a marker for HSC asymmetric and symmetric commitment. Overall, our results indicate that RNA methylation controls symmetric commitment and cell identity of HSCs and may provide a general mechanism for how stem cells regulate differentiation fate choice.
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Pricing and availability is not currently available.