Merck
  • MAPS: Model-based analysis of long-range chromatin interactions from PLAC-seq and HiChIP experiments.
MAPS: Model-based analysis of long-range chromatin interactions from PLAC-seq and HiChIP experiments.
PLoS computational biology (2019-04-16)
Ivan Juric, Miao Yu, Armen Abnousi, Ramya Raviram, Rongxin Fang, Yuan Zhao, Yanxiao Zhang, Yunjiang Qiu, Yuchen Yang, Yun Li, Bing Ren, Ming Hu
ABSTRACT
Hi-C and chromatin immunoprecipitation (ChIP) have been combined to identify long-range chromatin interactions genome-wide at reduced cost and enhanced resolution, but extracting information from the resulting datasets has been challenging. Here we describe a computational method, MAPS, Model-based Analysis of PLAC-seq and HiChIP, to process the data from such experiments and identify long-range chromatin interactions. MAPS adopts a zero-truncated Poisson regression framework to explicitly remove systematic biases in the PLAC-seq and HiChIP datasets, and then uses the normalized chromatin contact frequencies to identify significant chromatin interactions anchored at genomic regions bound by the protein of interest. MAPS shows superior performance over existing software tools in the analysis of chromatin interactions from multiple PLAC-seq and HiChIP datasets centered on different transcriptional factors and histone marks. MAPS is freely available at https://github.com/ijuric/MAPS.
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys4) Antibody, clone MC315, rabbit monoclonal, culture supernatant, clone MC315, Upstate®
Pricing and availability is not currently available.