Merck
  • Salmonella Translocated Effectors Recruit OSBP1 to the Phagosome to Promote Vacuolar Membrane Integrity.
Salmonella Translocated Effectors Recruit OSBP1 to the Phagosome to Promote Vacuolar Membrane Integrity.
Cell reports (2019-05-16)
Anna M Kolodziejek, Melissa A Altura, Junping Fan, Erik M Petersen, Matthew Cook, Peter S Brzovic, Samuel I Miller
ABSTRACT
Intracellular Salmonella use a type III secretion system (TTSS) to translocate effector proteins across the phagosome membrane and thus promote vacuole membrane tubulation, resulting in intracellular survival. This work demonstrates that the effector SseJ binds the eukaryotic lipid transporter oxysterol binding protein 1 (OSBP1). SseJ directs OSBP1 to the endosomal compartment in a manner dependent on the TTSS located on Salmonella pathogenicity island 2 (SPI2). OSBP1 localization is mediated by both SseJ and another OSBP1-binding SPI2 translocated effector, the deubiquitinase SseL. Deletion of both SseJ and SseL reduced vacuolar integrity with increased bacteria released into the eukaryotic cytoplasm of epithelial cells, indicating that their combined activities are necessary for vacuole membrane stability. Cells knocked down for OSBP1 or deleted for the OSBP1-binding proteins VAPA/B also demonstrate loss of vacuole integrity, consistent with the hypothesis that OSBP1 recruitment is required for SPI2-mediated alterations that promote vacuolar integrity of salmonellae.
Sigma-Aldrich
Anti-VAPA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-VAPB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-OSBP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.