Merck
  • ACSS2 promotes systemic fat storage and utilization through selective regulation of genes involved in lipid metabolism.
ACSS2 promotes systemic fat storage and utilization through selective regulation of genes involved in lipid metabolism.
Proceedings of the National Academy of Sciences of the United States of America (2018-09-20)
Zhiguang Huang, Menglu Zhang, Abigail A Plec, Sandi Jo Estill, Ling Cai, Joyce J Repa, Steven L McKnight, Benjamin P Tu
ABSTRACT
Acetyl-CoA synthetase 2 (ACSS2) is a conserved nucleocytosolic enzyme that converts acetate to acetyl-CoA. Adult mice lacking ACSS2 appear phenotypically normal but exhibit reduced tumor burdens in mouse models of liver cancer. The normal physiological functions of this alternate pathway of acetyl-CoA synthesis remain unclear, however. Here, we reveal that mice lacking ACSS2 exhibit a significant reduction in body weight and hepatic steatosis in a diet-induced obesity model. ACSS2 deficiency reduces dietary lipid absorption by the intestine and also perturbs repartitioning and utilization of triglycerides from adipose tissue to the liver due to lowered expression of lipid transporters and fatty acid oxidation genes. In this manner, ACSS2 promotes the systemic storage or metabolism of fat according to the fed or fasted state through the selective regulation of genes involved in lipid metabolism. Thus, targeting ACSS2 may offer a therapeutic benefit for the treatment of fatty liver disease.
Sigma-Aldrich
Anti-FABP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.