Merck
  • Interleukin-15 facilitates muscle regeneration through modulation of fibro/adipogenic progenitors.
Interleukin-15 facilitates muscle regeneration through modulation of fibro/adipogenic progenitors.
Cell communication and signaling : CCS (2018-07-22)
Xia Kang, Ming-Yu Yang, You-Xing Shi, Mei-Ming Xie, Min Zhu, Xiao-Long Zheng, Chen-Ke Zhang, Zi-Lu Ge, Xu-Ting Bian, Jing-Tong Lv, Yun-Jiao Wang, Bing-Hua Zhou, Kang-Lai Tang
ABSTRACT
Chronic muscle injury is characteristics of fatty infiltration and fibrosis. Recently, fibro/adipogenic progenitors (FAPs) were found to be indispensable for muscular regeneration while were also responsible for fibrosis and fatty infiltration in muscle injury. Many myokines have been proven to regulate the adipose or cell proliferation. Because the fate of FAPs is largely dependent on microenvironment and the regulation of myokines on FAPs is still unclear. We screened the potential myokines and found Interleukin-15 (IL-15) may regulate the fatty infiltration in muscle injury. In this study, we investigated how IL-15 regulated FAPs in muscle injury and the effect on muscle regeneration. Cell proliferation assay, western blots, qRT-PCR, immunohistochemistry, flow cytometric analysis were performed to investigate the effect of IL-15 on proliferation and adipogensis of FAPs. Acute muscle injury was induced by injection of glycerol or cardiotoxin to analyze how IL-15 effected on FAPs in vivo and its function on fatty infiltration or muscle regeneration. We identified that the expression of IL-15 in injured muscle was negatively associated with fatty infiltration. IL-15 can stimulate the proliferation of FAPs and prevent the adipogenesis of FAPs in vitro and in vivo. The growth of FAPs caused by IL-15 was mediated through JAK-STAT pathway. In addition, desert hedgehog pathway may participate in IL-15 inhibiting adipogenesis of FAPs. Our study showed IL-15 can cause the fibrosis after muscle damage and promote the myofiber regeneration. Finally, the expression of IL-15 was positively associated with severity of fibrosis and number of FAPs in patients with chronic rotator cuff tear. These findings supported the potential role of IL-15 as a modulator on fate of FAPs in injured muscle and as a novel therapy for chronic muscle injury.
Sigma-Aldrich
SAR-20347, ≥98% (HPLC)
Pricing and availability is not currently available.