Merck
Detecting CO2-Sensitive Hemichannels in Neurons in Acute Brain Slices.
STAR protocols (2020-12-31)
Emily Hill, Nicholas Dale, Mark J Wall
ABSTRACT
This protocol provides two independent methods to functionally detect the neuronal expression of CO2-sensitive hemichannels. These hemichannels (consisting of connexins 26 or 30) are directly gated by CO2, independent of pH changes and until recently were thought to be only expressed by glia. This protocol outlines a method to change the concentration of CO2 without changing pH, using isohydric solutions and then utilizing this to detect opening and closing of functional hemichannels using whole-cell patch clamp recording and dye loading. For complete details on the use and execution of this protocol, please refer to Hill et al. (2020).
Sigma-Aldrich
Phosphocreatine disodium salt hydrate, ≥97%
Sigma-Aldrich
Guanosine 5′-triphosphate tris salt, ≥93% (HPLC), powder
Sigma-Aldrich
Potassium D-gluconate, ≥99% (HPLC)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Adenosine 5′-triphosphate magnesium salt, ≥95%, bacterial
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture