Merck
  • Synthesis and antitubercular evaluation of amidoalkyl dibenzofuranols and 1H-benzo[2,3]benzofuro[4,5-e][1,3]oxazin-3(2H)-ones.
Synthesis and antitubercular evaluation of amidoalkyl dibenzofuranols and 1H-benzo[2,3]benzofuro[4,5-e][1,3]oxazin-3(2H)-ones.
Bioorganic & medicinal chemistry letters (2011-06-15)
Srinivas Kantevari, Thirumal Yempala, Perumal Yogeeswari, Dharmarajan Sriram, Balasubramanian Sridhar
ABSTRACT
A new class of amidoalkyl dibenzofuranols and 1H-benzo[2,3]benzofuro[4,5-e][1,3]oxazin-3(2H)-ones was synthesized in very good yields through polyphosphoric acid supported on silica (PPA-SiO(2)) catalyzed one-pot three component condensation of 2-dibenzofuranol; aromatic aldehydes and acetamide or benzamide or urea under solvent free conditions. At 125°C the reaction led to the formation of amidoalkyl dibenzofuranols 5a-k where as at 160°C cyclization take place to give oxazin-3(2H)-one analogues 6a-e. Screening all the 16 compounds for in vitro antimycobacterial activity against Mycobacterium tuberculosis H37Rv (MTB) resulted 1-((4-chlorophenyl)(2-hydroxydibenzo[b,d]furanyl)methyl)urea 5h; 1-((4-bromophenyl)(2-hydroxydibenzo[b,d]furanyl)methyl)urea 5i; 1-phenyl-1H-benzo[2,3]benzo furo[4,5-e][1,3]oxazin-3(2H)-one 6a (MIC 3.13μg/mL) and 1-(4-chlorophenyl)-1H-benzo[2,3]benzofuro[4,5-e][1,3]oxazin-3(2H)-one 6b; 1-(4-bromophenyl)-1H-benzo[2,3]benzofuro [4,5-e][1,3]oxazin-3(2H)-one 6c (MIC 1.56μg/mL) as most active antitubercular agents.
Sigma-Aldrich
Polyphosphoric acid, reagent grade, 115% H3PO4 basis
Sigma-Aldrich
Pyrazinecarboxamide
Sigma-Aldrich
Polyphosphoric acid, ~105% H3PO4 basis
Supelco
Isoniazid, analytical standard, ≥99% (TLC)