Merck
Genome engineering using CRISPR-Cas9 system.
Methods in molecular biology (Clifton, N.J.) (2014-11-20)
Le Cong, Feng Zhang
ABSTRACT
The Clustered Regularly Interspaced Short Palindromic Repeats (CRISPR)-Cas9 system is an adaptive immune system that exists in a variety of microbes. It could be engineered to function in eukaryotic cells as a fast, low-cost, efficient, and scalable tool for manipulating genomic sequences. In this chapter, detailed protocols are described for harnessing the CRISPR-Cas9 system from Streptococcus pyogenes to enable RNA-guided genome engineering applications in mammalian cells. We present all relevant methods including the initial site selection, molecular cloning, delivery of guide RNAs (gRNAs) and Cas9 into mammalian cells, verification of target cleavage, and assays for detecting genomic modification including indels and homologous recombination. These tools provide researchers with new instruments that accelerate both forward and reverse genetics efforts.
Sigma-Aldrich
ROCK Inhibitor (Y-27632), The ROCK Inhibitor (Y-27632) is available in a 5 mg format & has been optimized & validated for cell culture & Neuroscience applications.
Pricing and availability is not currently available.
Sigma-Aldrich
LB Broth with agar (Lennox), Highly-referenced microbial growth powder medium with Agar, low salt, suitable for salt-sensitive E. coli culture.
Pricing and availability is not currently available.
Sigma-Aldrich
LB Broth (Lennox), Highly-referenced microbial growth powder medium, low salt, suitable for salt-sensitive E.coli culture.
Pricing and availability is not currently available.