Merck
  • Overactivation of the androgen receptor exacerbates gravid uterine ferroptosis via interaction with and suppression of the NRF2 defense signaling pathway.
Overactivation of the androgen receptor exacerbates gravid uterine ferroptosis via interaction with and suppression of the NRF2 defense signaling pathway.
FEBS letters (2022-01-18)
Min Hu, Yuehui Zhang, Lingjing Lu, Yu Zhou, Denghui Wu, Mats Brännström, Linus R Shao, Håkan Billig
ABSTRACT
The mechanisms through which the androgen-dependent activation of the androgen receptor (AR) regulates gravid uterine ferroptosis remain unknown. We show that while co-exposure of pregnant rats to the androgen 5α-dihydrotestosterone (DHT) and insulin (INS) triggered uterine ferroptotic signaling cascades, additional treatment with the anti-androgen flutamide increased expression of the key ferroptosis-inhibitory proteins SLC7A11, GSH, and GPX4; reduced iron content; normalized levels of ferroptosis-associated Tfrc, Fpn1, and Ho1 mRNAs; reduced levels of proteins modified by 4-HNE (a marker of ferroptosis); and restored protein levels of NRF2, a key transcription factor regulating antioxidant defense signaling, in the gravid uterus. Furthermore, exposure to DHT alone increased uterine ferroptosis, and NRF2 abundance was negatively correlated with AR status. Co-immunoprecipitation and Western blot assays revealed that the AR physically interacted with endogenous NRF2, and this interaction was increased by DHT exposure in vivo. Our results suggest that AR overactivation and NRF2 suppression cooperate in the regulation of NRF2-targets in uterine ferroptosis.
Product Description
Millipore
Protein G Plus/Protein A Agarose Suspension, Protein G PLUS/Protein A-Agarose mixture specifically formulated for immunoprecipitation.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture