Merck
  • Recombinant human tumor necrosis factor-alpha: effects on proliferation of normal and transformed cells in vitro.
Recombinant human tumor necrosis factor-alpha: effects on proliferation of normal and transformed cells in vitro.
Science (New York, N.Y.) (1985-11-22)
B J Sugarman, B B Aggarwal, P E Hass, I S Figari, M A Palladino, H M Shepard
ABSTRACT
Modulation of the growth of human and murine cell lines in vitro by recombinant human tumor necrosis factor-alpha (rTNF-alpha) and recombinant human interferon-gamma (rIFN-gamma) was investigated. rTNF-alpha had cytostatic or cytolytic effects on only some tumor cell lines. When administered together with rIFN-gamma, rTNF-alpha showed enhanced antiproliferative effects on a subset of the cell lines tested. In contrast to its effects on sensitive tumor cells, rTNF-alpha augmented the growth of normal diploid fibroblasts. Variations in the proliferative response induced by rTNF-alpha were apparently not due to differences in either the number of binding sites per cell or their affinity for rTNF-alpha. These observations indicate that the effects of rTNF-alpha on cell growth are not limited to tumor cells, but rather that this protein may have a broad spectrum of activities in vivo.
Product Description
WiDr, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Pricing and availability is not currently available.