Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, suitable for protein sequencing, salt-free, lyophilized powder
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, ≥40 units/mg protein, vial of 5 mg
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type I-S, essentially salt-free, lyophilized powder
Sigma-Aldrich
Trypsin from porcine pancreas, Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, Type II-S, 1,000-2,000 units/mg dry solid
Sigma-Aldrich
α-Chymotrypsin−Agarose from bovine pancreas, lyophilized powder, 2,000-3,500 units/g agarose (One ml gel will yield 65-120 units)
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, BioReagent, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
Trypsin from porcine pancreas, tablet, 1 mg tablet
Sigma-Aldrich
α-Chymotrypsin from human pancreas, lyophilized powder
Sigma-Aldrich
Trypsin from bovine pancreas, suitable for protein sequencing, lyophilized powder
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type II, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
TrypZean® bovine, recombinant, expressed in corn, lyophilized powder, ≥3,350 units/mg solid (USP)
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, (TLCK treated to inactivate residual tryspin activity), Type VII, essentially salt-free, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
Trypsin from porcine pancreas, ~1500 U/mg
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from human pancreas, salt-free, lyophilized powder, vial of ≥1,000 BAEE units
Sigma-Aldrich
Trypsin Acetylated from bovine pancreas, Type V-S, ≥8,500 BAEE units/mg protein (biuret)
Sigma-Aldrich
Trypsin from bovine pancreas, essentially salt-free, lyophilized powder, ≥9,000 BAEE units/mg protein, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypsin from bovine pancreas, Type I, ~10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, Type XI, lyophilized powder, ≥6,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, ≥2,500 USP units/mg solid, meets USP testing specifications
Sigma-Aldrich
Trypsin from bovine pancreas, powder, ≥7,500 BAEE units/mg solid