Merck
  • [Clinical effectiveness and tolerance of climbazole containing dandruff shampoo in patients with seborrheic scalp eczema].
[Clinical effectiveness and tolerance of climbazole containing dandruff shampoo in patients with seborrheic scalp eczema].
Praxis (2001-09-06)
W Wigger-Alberti, K Kluge, P Elsner
ABSTRACT
Pityrosporum ovale appears to play an important role in the pathogenesis of dandruff as a symptom of seborrheic dermatitis. Climbazole is an antimycotic agent with a high in vitro and in vivo efficacy against P. ovale. In the presented work we investigated the efficacy and safety of a climbazole 0.65% shampoo on seborrheic dermatitis of 30 volunteers. Subjects were diagnosed as having moderate to severe seborrheic dermatitis of the scalp. After a 1-week washout and a 4-week treatment the clinical evaluation showed a successful reduction of dandruff, skin redness and itching in 80% of the volunteers and a mild improvement in 20% of the volunteers. The cosmetic acceptability was very good by the majority. It is concluded that the formulation tested is effective in the treatment of moderate to severe dandruff.
Product Description
Supelco
Climbazole, PESTANAL®, analytical standard
Pricing and availability is not currently available.