Merck
  • Photodegradation of fluoroquinolones in surface water and antimicrobial activity of the photoproducts.
Photodegradation of fluoroquinolones in surface water and antimicrobial activity of the photoproducts.
Water research (2012-08-21)
Michela Sturini, Andrea Speltini, Federica Maraschi, Luca Pretali, Antonella Profumo, Elisa Fasani, Angelo Albini, Roberta Migliavacca, Elisabetta Nucleo
ABSTRACT
The widespread presence of fluoroquinolone antibiotics (FQs) in natural ecosystems is a health hazard for humans and other living organisms. The role of sunlight in degrading FQs present in environmental waters has been studied. In particular, the photodegradation of four largely employed FQs, viz. Ciprofloxacin (CIP), Danofloxacin (DAN), Levofloxacin (LEV) and Moxifloxacin (MOX) has been studied in not tampered river water. Degradation rates have been investigated at ppb levels (20-50 μg L(-1)) under solar light, and the results have been commented critically. The products distribution has been studied by HPLC-ESI-MS/MS analysis and structures have been attributed on the basis of their mass fragmentation spectra. Importantly from the environmental point of view, the (potentially toxic) FQ nucleus remained intact over the early stages of the degradation. Indeed, the photoproducts were proved to possess residual antibacterial activity, as shown from in vitro antibacterial activity tests against different well characterized human and environmental bacterial strains, carried out on the above FQs, as well as for Enrofloxacin (ENR) and Marbofloxacin (MAR).
Product Description
Supelco
Danofloxacin, VETRANAL®, analytical standard
Pricing and availability is not currently available.