Merck
Common trace elements alleviate pain in an experimental mouse model.
Journal of neuroscience research (2013-01-31)
Bogdan I Tamba, Maria-Magdalena Leon, Tudor Petreus
ABSTRACT
Trace elements represent a group of essential metals or metaloids necessary for life, present in minute amounts. Analgesic adjuvants can enhance the effect of other pain drugs or be used for pain control themselves. Previous studies on the effects of trace elements on nociception and their potential use as analgesic adjuvants have yielded conflicting results. In this study, we tested the hypothesis that three vital trace elements (Zn²⁺, Mg²⁺, Cu²⁺) have direct antinociceptive effects. Groups of eight Swiss mice were intraperitoneally (i.p) injected with incremental concentrations of Zn²⁺ sulfate (0.5, 2.0 mg/kg), Zn²⁺ citrate (0.125, 0.5 mg/kg), Mg²⁺ chloride (37.5, 75, 150 mg/kg), Cu²⁺ chloride (0.5, 1.0, 2.0 mg/kg), and Cu²⁺ sulfate (0.5, 1.0 mg/kg) or saline (control). Evaluations were made by hot plate (HP) and tail flick (TF) tests for central antinociceptive effect, writhing test (WT) for visceral antinociceptive effect, and activity cage (AC) test for spontaneous behavior. Zn²⁺ induced pain inhibition in HP/TF tests (up to 17%) and WT (up to 25%), with no significant differences among the salts used. Mg²⁺ salts induced pain inhibition for all performed tests (up to 85% in WT). Cu²⁺ salts showed antinociceptive effects for HP/TF (up to 28.6%) and WT (57.28%). Only Mg²⁺ and Cu²⁺ salts have displayed significant effects in AC (Mg²⁺ anxiolytic/depressant effect; Cu²⁺ anxiolytic effect). We interpret these data to mean that all tested trace elements induced antinociceptive effects in central and visceral pain tests. Our data indicate the potential use of these cheap adjuvants in pain therapy.
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Pricing and availability is not currently available.
Sigma-Aldrich
Copper(II) chloride dihydrate, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Copper(II) chloride dihydrate, reagent grade
Pricing and availability is not currently available.
Sigma-Aldrich
Copper(II) chloride dihydrate, powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Copper(II) sulfate, anhydrous, powder, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Copper(II) chloride dihydrate, ≥99.95% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Copper(II) sulfate, ReagentPlus®, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Copper(II) sulfate solution, 4 % (w/v) (prepared from copper (II) sulfate pentahydrate)
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Pricing and availability is not currently available.
Sigma-Aldrich
Copper(II) chloride dihydrate, 99.999%
Pricing and availability is not currently available.
Sigma-Aldrich
Zinc sulfate heptahydrate, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Zinc sulfate heptahydrate, ReagentPlus®, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Zinc sulfate heptahydrate, ≥99.95% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Zinc sulfate heptahydrate, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Zinc sulfate heptahydrate, suitable for plant cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Zinc sulfate heptahydrate, ACS reagent, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Zinc sulfate heptahydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Zinc sulfate solution, BioUltra, Molecular Biology, 2.0 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Zinc sulfate solution, 0.3 N
Pricing and availability is not currently available.
Sigma-Aldrich
Copper(II) chloride, anhydrous, powder, ≥99.995% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Copper(II) chloride, 99.999% trace metals basis
Pricing and availability is not currently available.