Merck
  • Sodium dodecyl sulphate, a strong inducer of thermostable glucanhydrolase secretion from a derepressed mutant strain of Bacillus alcalophilus GCBNA-4.
Sodium dodecyl sulphate, a strong inducer of thermostable glucanhydrolase secretion from a derepressed mutant strain of Bacillus alcalophilus GCBNA-4.
Applied biochemistry and biotechnology (2013-03-05)
Nadia Shamim, Sikander Ali, Ikram Ul-Haq
ABSTRACT
In the present study, we report the optimisation of batch conditions for improved α-1,4-glucan-glucanohydrolase (GGH) secretion by a nitrous acid (NA)-treated Bacillus alcalophilus. The wild (isolate GCB-18) and NA-derivative (mutant GCBNA-4) were grown in a medium containing 10 g/L nutrient broth, 10 g/L starch, 5 g/L lactose, 2 g/L ammonium sulphate, 2 g/L CaCl2 and phosphate buffer (pH 7.6). Sodium dodecyl sulphate (SDS) was used as an enzyme inducer while batch fermentations were carried out at 40 °C. The mutant produced GGH in 40 h which was 15-fold higher than the wild in presence of SDS. Thermodynamic studies revealed that the mutant culture exhibited the capability for improved enzyme activity over a broad range of temperature (35-70 °C). The enzyme was purified by cation-exchange column chromatography with ~80 % recovery. The performance of fuzzy-logic system control was found to be highly promising for the improved substrate conversion rate. The correlation (1.045E + 0025) among variables demonstrated the model terms as highly significant indicating commercial utility of the culture used (P < 0.05).
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O