Merck
  • Chemomechanical caries removal in primary molars : evaluation of marginal leakage and shear bond strength in bonded restorations--an in vitro study.
Chemomechanical caries removal in primary molars : evaluation of marginal leakage and shear bond strength in bonded restorations--an in vitro study.
The Journal of clinical pediatric dentistry (2013-07-17)
P M Viral, C Nagarathna, B S Shakuntala
ABSTRACT
To evaluate and compare the efficiency, marginal leakage and shear bond strength of Carisolv and Papacarie in primary molars. Freshly extracted 60 human carious primary molars were randomly divided into two experimental groups--Group I [caries removal by Carisolv] and Group II [caries removal by Papacarie]. The amount of time taken for complete caries removal was recorded using a stopwatch. After bonded restorations, both the experimental groups were further randomly subdivided into four experimental groups and subjected to marginal leakage and shear bond strength evaluation. Papacarie [337.67 +/- 18.13] was clinically more efficient than Carisolv [461.33 +/- 27.76] in removing caries with respect to time in seconds. 66.7% of teeth treated with Carisolv did not show any marginal leakage as compared to 20% with Papacarie. The mean [ +/- SD] shear bond strength of Carisolv [9.67 +/- 3.80] treated teeth was slightly more when compared to Papacarie [8.36 +/- 4.51]. Papacarie was clinically more efficient in caries removal but showed significantly more marginal leakage than Carisolv.
Product Description
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
L-Lysine acetate salt, ≥98% (HPLC)
Sigma-Aldrich
Papain from papaya latex, crude powder, 1.5-10 units/mg solid
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
L-Glutamic acid hydrochloride, ≥99% (HPLC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Papain from papaya latex, lyophilized powder, aseptically filled
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
L-Leucine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Leucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Papain from Carica papaya, solution, light brown, ≥10 U/mg protein (~25 mg/ml)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
L-Leucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
L-Leucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
L-Leucine, 99%, FG
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
L-Lysine, ≥98% (TLC)