Merck
  • Design of a new dental adhesive--effect of a water-soluble sodium acylphosphine oxide with crown ether on adhesion to dental hard tissues.
Design of a new dental adhesive--effect of a water-soluble sodium acylphosphine oxide with crown ether on adhesion to dental hard tissues.
Dental materials journal (2009-08-11)
Kunio Ikemura, Kensuke Ichizawa, Kiyomi Fuchigami, So Ito, Takeshi Endo
ABSTRACT
The behavior of water-soluble photoinitiators with crown ethers in dental adhesives is unknown. This study investigated the effect of sodium acylphosphine oxide (APO-Na) with crown ether in a hydrophobic adhesive on adhesion to teeth. Sodium 2,4,6-trimethylbenzoyl-phenylphosphine oxide (TMPO-Na = APO-Na) was synthesized in 67.1% yield and identified by 1H NMR. APO-Na was dissolved in hydrophobic resins in the presence of a crown ether (ionophore effect). Thirty kinds of experimental single-step adhesives comprising APO-Na, CE, Bis-GMA, 6-methacryloyloxyhexyl phosphonoacetate (6-MHPA), and 4-methacryloyloxyethyl trimellitic acid (4-MET) were thereby prepared. Shear bond strength to unetched ground teeth was measured at a crosshead speed of 1.0 mm/min, and the data were analyzed by ANOVA. The shear bond strength results of bonding resins containing APO-Na with 18-crown-6-ether (CE-6) were significantly higher than that without CE-6 (control) (p<0.05). Higher bond strength values [for enamel: BR24 at 19.3 (3.2) MPa; for dentin: BR29 at 20.2 (4.7) MPa] were achieved with the adhesives containing APO-Na, CE-6, 6-MHPA, and 4-MET. Therefore, it was found that APO-Na with CE-6 contributed to the efficient bonding performance of single-step adhesive to teeth. However, in view of the biosafety hazard posed by crown ethers, the search is still on for reagents that are biologically safer than crown ethers--but with ionophor effects--to be used in dental adhesives.
Sigma-Aldrich
Phenylphosphinic acid, 99%
Pricing and availability is not currently available.