Merck
  • Cellulosic binder-assisted formation of graphene-paper electrode with flat surface and porous internal structure.
Cellulosic binder-assisted formation of graphene-paper electrode with flat surface and porous internal structure.
Journal of nanoscience and nanotechnology (2013-11-20)
Chang Kee Lee, Sang Bong Lee, Sung Wook Hwang, Kwan Woo Park, Jin Kie Shim
ABSTRACT
We have reported the fabrication of flexible graphene-paper electrode (GPE) with a flat surface, whose internal structure has been formed with gradient porous build-up (from the surface to the 2-hydroxyethyl cellulose (HC)-coated paper). HC solution was used as a binder to form the gradient porous graphene layer, enabling it to create an anchoring force between the porous graphene layer and the filter paper. The morphology of GPE was investigated using a scanning electron microscope, and the surface resistance of the GPE as a function of graphene content was determined using four-probe method. The electrochemical performance of the GPE was evaluated using a three-electrode test cell by cyclic voltammetry. The gravimetric capacitance of GPE was found to be 120 F per gram of graphene, and the capacitance retention was within ca. 96% for over 500 cycles. This could be attributed to both the low surface resistance resulting from the flat surface and the high electrochemical activity caused by the gradient porous structure. This unique structure not only offers an enhanced conductivity and good electrical contact between the electrode and electrolyte but also helps GPE to maintain good cyclic stability, proving its potential for use in various rechargeable and portable energy-storage devices.
Product Description
Supelco
Avicel® PH-101, ~50 μm particle size
Pricing and availability is not currently available.
Sigma-Aldrich
Graphite, flakes
Pricing and availability is not currently available.
Sigma-Aldrich
Graphite, powder, <150 μm, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 6 mm, 99.995% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Graphite, powder, <45 μm, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 3 mm, low density, 99.995% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Graphite, powder, <20 μm, synthetic
Pricing and availability is not currently available.
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Pricing and availability is not currently available.
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Pricing and availability is not currently available.
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Pricing and availability is not currently available.
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
α-Cellulose, powder
Pricing and availability is not currently available.
Supelco
Cellulose, DS-0, powder, suitable for thin layer chromatography (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Cellulose, fibers, (medium)
Pricing and availability is not currently available.
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm
Pricing and availability is not currently available.
Supelco
Cellulose, acid washed, from spruce, suitable for column chromatography
Pricing and availability is not currently available.
Sigma-Aldrich
Cellulose, colloidal, microcrystalline
Pricing and availability is not currently available.
Sigma-Aldrich
Cellulose, microcrystalline, powder
Pricing and availability is not currently available.
Supelco
Cellulose, powder, suitable for column chromatography
Pricing and availability is not currently available.
Supelco
Cellulose, acid washed, powder, suitable for column chromatography
Pricing and availability is not currently available.
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Pricing and availability is not currently available.
Supelco
Cellulose, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Pricing and availability is not currently available.