Merck
  • Involvement of alanine racemase in germination of Bacillus cereus spores lacking an intact exosporium.
Involvement of alanine racemase in germination of Bacillus cereus spores lacking an intact exosporium.
Archives of microbiology (2013-12-19)
Elena Venir, Manuela Del Torre, Vincenzo Cunsolo, Rosaria Saletti, Rita Musetti, Mara Lucia Stecchini
ABSTRACT
The L-alanine mediated germination of food isolated Bacillus cereus DSA 1 spores, which lacked an intact exosporium, increased in the presence of D-cycloserine (DCS), which is an alanine racemase (Alr) inhibitor, reflecting the activity of the Alr enzyme, capable of converting L-alanine to the germination inhibitor D-alanine. Proteomic analysis of the alkaline extracts of the spore proteins, which include exosporium and coat proteins, confirmed that Alr was present in the B. cereus DSA 1 spores and matched to that encoded by B. cereus ATCC 14579, whose spore germination was strongly affected by the block of conversion of L- to D-alanine. Unlike ATCC 14579 spores, L-alanine germination of B. cereus DSA 1 spores was not affected by the preincubation with DCS, suggesting a lack of restriction in the reactant accessibility.
Sigma-Aldrich
L-Alanine, ≥99%
Pricing and availability is not currently available.
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Pricing and availability is not currently available.
Alanine, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
L-Alanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Sigma-Aldrich
D-Cycloserine
Pricing and availability is not currently available.
Sigma-Aldrich
L-Alanine-12C3, 99.9 atom % 12C
Pricing and availability is not currently available.
Sigma-Aldrich
D-Cycloserine
Pricing and availability is not currently available.
Sigma-Aldrich
D-Cycloserine, synthetic
Pricing and availability is not currently available.