Merck
  • The effects of general anaesthesia on memory in children: a comparison between propofol and sevoflurane.
The effects of general anaesthesia on memory in children: a comparison between propofol and sevoflurane.
Anaesthesia (2014-01-22)
J Yin, S-L Wang, X-B Liu
ABSTRACT
We studied the effects of general anaesthesia on memory 7 days and 3 months following elective hernia surgery. Sixty children aged between 7 and 13 years were randomly allocated to receive either propofol or sevoflurane. Memory was classified into immediate, short-term and long-term memory and assessed using the Wechsler Memory Scale-Propofol impaired short-term memory 7 days postoperatively compared with pre-operative values (image recalling: p = 0.02, figure recognition: p = 0.01, visual reproduction: p = 0.03) but recovered to baseline levels 3 months following surgery. Neither general anaesthetic affected immediate or long-term memory. We conclude that propofol impairs short-term memory postoperatively in children.
Sigma-Aldrich
2,6-Diisopropylphenol, 97%
Supelco
Propofol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sevoflurane, European Pharmacopoeia (EP) Reference Standard
Propofol for peak identification, European Pharmacopoeia (EP) Reference Standard
Propofol, European Pharmacopoeia (EP) Reference Standard