Merck
  • Circulating microRNA profiles altered in mice after 28 d exposure to perfluorooctanoic acid.
Circulating microRNA profiles altered in mice after 28 d exposure to perfluorooctanoic acid.
Toxicology letters (2014-01-25)
Shengmin Yan, Jianshe Wang, Wei Zhang, Jiayin Dai
ABSTRACT
Perfluorooctanoic acid (PFOA) is a stable man-made compound with many industrial and commercial uses. Recently, however, concern has been raised that it may induce various toxicological effects such as hepatotoxicity, immunotoxicity, and developmental toxicity. Because levels of circulating microRNAs (miRNAs) can be altered in several clinical diseases, they may serve as potential novel biomarkers. Here,we explored differences in the profiles of circulating miRNAs in mice after PFOA exposure. Using TaqMan miRNA arrays, we determined that the levels of 24 circulating miRNAs were altered in mice dosed with PFOA at 1.25 mg/kg/d and 73 were altered in mice dosed with 5 mg/kg/d. Eight miRNAs were further validated using TaqMan Real-Time PCR assays. Results were consistent with those obtained from the TaqMan miRNA arrays, except for miR-199a-3p. The most remarkable of the circulating miRNAs (miR-26b-5p and miR-199a-3p) were also up-regulated in the serum of occupational workers in our previous epidemiological study. We also found similar patterns in mice exposed to PFOS. These results demon-strated that circulating miRNA profiles were altered after exposure to high concentrations of PFOA and miR-28-5p, miR-32-5p, miR-122-5p, miR-192-5p, and miR-26b-5p in serum may be linked to effects of PFOA, especially in occupationally exposed people.
Sigma-Aldrich
Pentadecafluorooctanoic acid ammonium salt, ≥98.0% (sum of isomers)
Sigma-Aldrich
Perfluorooctanoic acid, 95%
Supelco
Pentadecafluorooctanoic acid solution, 100 μg/mL in methanol, analytical standard