Merck
  • Inhibition of TNF-alpha, IL-1beta, and IL-6 productions and NF-kappa B activation in lipopolysaccharide-activated RAW 264.7 macrophages by catalposide, an iridoid glycoside isolated from Catalpa ovata G. Don (Bignoniaceae).
Inhibition of TNF-alpha, IL-1beta, and IL-6 productions and NF-kappa B activation in lipopolysaccharide-activated RAW 264.7 macrophages by catalposide, an iridoid glycoside isolated from Catalpa ovata G. Don (Bignoniaceae).
International immunopharmacology (2002-09-28)
S J An, H O Pae, G S Oh, B M Choi, S Jeong, S I Jang, H Oh, T O Kwon, C E Song, H T Chung
ABSTRACT
Catalposide, the major iridoid glycoside isolated from the stem bark of Catalpa ovata G. Don (Bignoniaceae), was found to inhibit the productions of tumor necrosis factor-alpha (TNF-alpha), interleukin-1beta (IL-1beta), and interleukin-6 (IL-6), and the activation of nuclear factor kappaB (NF-kappaB) in RAW 264.7 macrophages activated with lipopolysaccharide (LPS). Catalposide also inhibited the expressions of TNF-alpha, IL-1beta, and IL-6 genes and the nuclear translocation of p65 subunit of NF-kappaB in LPS-activated RAW 264.7 cells. Flow cytometric analysis revealed that catalposide suppressed the binding of FITC-conjugated LPS to CD14 on the surface of cells, probably resulting in the inhibitory effects on TNF-alpha, IL-1beta, and IL-6 productions and NF-kappaB activation. These findings suggest that catalposide could be an attractive candidate for adjunctive therapy in gram-negative bacterial infections.
Sigma-Aldrich
Catalposide, ≥95% (LC/MS-ELSD)
Pricing and availability is not currently available.