Merck
  • Design and in vitro evaluation of simvastatin-hydroxyapatite coatings by an electrochemical process on titanium surfaces.
Design and in vitro evaluation of simvastatin-hydroxyapatite coatings by an electrochemical process on titanium surfaces.
Journal of biomedical nanotechnology (2014-05-09)
Zhao Shifang, Shi Jue, He Fuming, Li Liu, Yang Guoli
ABSTRACT
The aim of this study was to deposit simvastatin-hydroxyapatite (sim-HA) coatings on titanium surfaces by an electrochemical process and evaluate osteoprogenitor cell responses to sim-HA-coated surfaces. Simvastatin was prepared onto titanium surfaces with varying concentration (10(-7), 10(-6), 10(-5) and 10(-4) mol/L). Surface characteristics were performed by FSEM, XRD and FTIR. LC-MS/MS method was used for simvastatin quantification in Sim-HA-coated surfaces. Murine calvaria-derived pre-osteoblastic cell (MC3T3-E1) proliferation, alkaline phosphatase activity (ALP) and osteocalcin release were used to measure osteoblastic activities. FSEM observation showed rod-like HA crystals covered on all surfaces. As drug concentration increased, the crystal diameter decreased. XRD and FTIR observations showed all coatings consisted of HA. LC-MS/MS test showed the simvastatin concentration in 10(-6) mol/L group was 2.77 x 10(-7) mol/L/cm2, while in 10(-7) mol/L group was 1.89 x 10(-7) mol/L/cm2. MC3T3-E1 cells grown on 10(-7) mol/L and 10(-6) mol/L Sim-HA surfaces showed increased ALP activity as compared to HA-coated surfaces on day 7 (P < 0.05) while at 10 day, all Sim-HA groups were significantly increased than HA group (P < 0.05). Two Sim-HA groups showed significant increases in osteocalcin production on 7 10 and 14 day (P < 0.05). It showed that incubation with 10(-7) mol/L simvastatin enhanced the cell proliferation to a statistically significant extent (P < 0.05) compared with the HA group at day 7. It was concluded that simvastatin was successfully deposited into HA coatings using the electrochemical process and the sim-HA coatings enhanced differentiation of osteoprogenitor cells.
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Titanium, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.995% trace metals basis
Supelco
Simvastatin, analytical standard
Titanium, IRMM®, certified reference material, 0.1 mm foil
Simvastatin, European Pharmacopoeia (EP) Reference Standard
Titanium, rod, 500mm, diameter 16mm, annealed, 99.6+%
Titanium, sphere, 200pcs, diameter 3mm, 99.6+%
Titanium, tube, 100mm, outside diameter 6.35mm, inside diameter 4.57mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, rod, 200mm, diameter 6mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 12.7mm, inside diameter 10.9mm, wall thickness 0.9mm, annealed, 99.6+%
Titanium, wire reel, 500m, diameter 0.25mm, as drawn, 99.6+%
Titanium, wire reel, 2m, diameter 0.5mm, as drawn, 99.99+%
Titanium, wire reel, 5m, diameter 0.5mm, as drawn, 99.99+%
Titanium, wire reel, 1m, diameter 0.75mm, as drawn, 99.6+%
Titanium, wire reel, 200m, diameter 0.05mm, hard, 99.8%
Titanium, wire reel, 1m, diameter 1.0mm, annealed, 99.6+%