Merck
  • Helicopter crashes into water: warning time, final position, and other factors affecting survival.
Helicopter crashes into water: warning time, final position, and other factors affecting survival.
Aviation, space, and environmental medicine (2014-04-24)
Christopher J Brooks, Conor V MacDonald, Susan P Baker, Dennis F Shanahan, Wren L Haaland
ABSTRACT
According to 40 yr of data, the fatality rate for a helicopter crash into water is approximately 25%. Does warning time and the final position of the helicopter in the water influence the survival rate? The National Transportation Safety Board (NTSB) database was queried to identify helicopter crashes into water between 1981 and 2011 in the Gulf of Mexico and Hawaii. Fatality rate, amount of warning time prior to the crash, and final position of the helicopter were identified. There were 133 helicopters that crashed into water with 456 crew and passengers. Of these, 119 occupants (26%) did not survive; of those who did survive, 38% were injured. Twelve died after making a successful escape from the helicopter. Crashes with < 15 s warning had a fatality rate of 22%, compared to 12% for 16-60 s warning and 5% for > 1 min. However, more than half of fatalities (57%) came from crashes for which the warning time could not be determined. Lack of warning time and how to survive in the water after the crash should be a topic for study in all marine survival/aircraft ditching courses. Investigators should be trained to provide estimates of warning time when investigating helicopter crashes into water.
Product Description
Supelco
Water, for TOC analysis
Supelco
Water
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
Water, for ultratrace analysis, ACS reagent
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O