Merck
  • The effectiveness of N-Acetylcysteine in preventing contrast-induced nephropathy in patients undergoing contrast-enhanced computed tomography: a meta-analysis of randomized controlled trials.
The effectiveness of N-Acetylcysteine in preventing contrast-induced nephropathy in patients undergoing contrast-enhanced computed tomography: a meta-analysis of randomized controlled trials.
International urology and nephrology (2013-01-04)
Mei-Yi Wu, Hui-Fen Hsiang, Chung-Shun Wong, Min-Szu Yao, Yun-Wen Li, Chao-Ying Hsiang, Chyi-Huey Bai, Yung-Ho Hsu, Yuh-Feng Lin, Ka-Wai Tam
ABSTRACT
N-Acetylcysteine (NAC) is reported to have potential for preventing of contrast-induced nephropathy (CIN) in patients undergoing coronary angiography. However, the effectiveness of NAC in preventing CIN in patients undergoing contrast-enhanced computed tomography (CT) is still controversial. We conducted a meta-analysis of relevant randomized controlled trials (RCTs) to further examine this issue. RCTs were identified by computerized searching in PubMed, EMBASE, SCOPUS, and Cochrane databases. Two reviewers independently assessed the methodological quality of each study. A meta-analysis was performed to evaluate the effectiveness of NAC in preventing CIN in patients undergoing CT. The primary outcome was the incidence of contrast-induced nephropathy, and the requirement for dialysis. The secondary outcome was the change of serum creatinine. Six randomized controlled trials were identified with a total of 496 patients meeting the criteria for this study. Prophylactic administration of NAC in patients with serum creatinine above 1.2 mg/dL undergoing contrast-enhanced CT, along with hydration, reduced the risk of CIN (relative risk 0.20; 95 % confidence interval: 0.07-0.57). Requirement for dialysis was not significantly different between the NAC group and the control group. This review provides evidence of the efficacy of NAC in preventing the incidence of CIN and recommends that NAC be more widely used in high-risk patients undergoing contrast-enhanced CT. On the basis of the evidence reviewed, further research involving large RCTs may be warranted.
Product Description
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
USP
Acetylcysteine, United States Pharmacopeia (USP) Reference Standard
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard