Merck
  • Apoptosis-inducing effects of Melissa officinalis L. essential oil in glioblastoma multiforme cells.
Apoptosis-inducing effects of Melissa officinalis L. essential oil in glioblastoma multiforme cells.
Cancer investigation (2014-04-22)
Rafaela Muniz de Queiroz, Christina Maeda Takiya, Lívia Paes Tavares Pacheco Guimarães, Gleice da Graça Rocha, Daniela Sales Alviano, Arie Fitzgerald Blank, Celuta Sales Alviano, Cerli Rocha Gattass
ABSTRACT
Current therapies for glioblastoma multiforme (GBM) are not effective. This study investigated the activity of the M. officinalis essential oil (EO) and its major component (citral) in GBM cell lines. Both EO and citral decreased the viability and induced apoptosis of GBM cells as demonstrated by DNA fragmentation and caspase-3 activation. Antioxidant prevented citral-induced death, indicating its dependence on the production of reactive oxygen species. Citral downmodulated the activity and inhibited the expression of multidrug resistance associated protein 1 (MRP1). These results show that EO, through its major component, citral, may be of potential interest for the treatment of GBM.
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
USP
Acetylcysteine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
Rhodamine 123, mitochondrial specific fluorescent dye
Sigma-Aldrich
DAPI, for nucleic acid staining
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Rhodamine 123, BioReagent, suitable for fluorescence, ≥85% (HPLC)
Sigma-Aldrich
Citral, natural, ≥96%, FCC, FG
Sigma-Aldrich
Citral, 95%
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
Citral, mixture of cis and trans, ≥96%, FG
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Supelco
Citral, analytical standard
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®